Assay Details
Target Gene Details
Entrez Gene ID: | 10526 |
Gene Name: | importin 8 |
Gene Aliases: |
RANBP8 |
Location: |
Chr.12:30628981-30695995 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IPO8 | NM_001190995.1 | 21 | 3997 | NP_001177924.1 |
| NM_006390.3 | 25 | 4635 | NP_006381.2 | |
| XM_017018691.1 | 25 | 4532 | XP_016874180.1 | |
| XM_017018692.1 | 24 | 4192 | XP_016874181.1 | |
| AB209602.1 | 11 | 3652 | BAD92839.1 | |
| AK000201.1 | 1 | 223 | ||
| AK025607.1 | 1 | 2066 | ||
| AK130090.1 | 5 | 1814 | ||
| BG574795.1 | ||||
| EB387074.1 | 1 | 668 |
More Information
Set Membership: |
|

Genomic Map