Assay Details
Target Gene Details
Entrez Gene ID: | 11143 |
Gene Name: | lysine acetyltransferase 7 |
Gene Aliases: |
HBO1, HBOA, MYST2, ZC2HC7 |
Location: |
Chr.17:49788619-49829096 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KAT7 | NM_001199155.1 | 3 | 462 | NP_001186084.1 |
| NM_001199156.1 | NP_001186085.1 | |||
| NM_001199157.1 | 3 | 462 | NP_001186086.1 | |
| NM_001199158.1 | NP_001186087.1 | |||
| NM_007067.4 | 3 | 462 | NP_008998.1 | |
| AF074606.1 | 3 | 288 | AAC99368.1 | |
| AF140360.1 | 3 | 385 | AAD42348.1 | |
| AF217502.1 | 3 | 370 | AAL56649.1 | |
| AK023890.1 | 3 | 393 | ||
| AK122932.1 | 3 | 828 | ||
| AK293976.1 | ||||
| AK294014.1 | 3 | 399 | ||
| AK294052.1 | ||||
| AK294836.1 | 4 | 367 | ||
| AK297212.1 | 3 | 544 | ||
| BC032640.2 | 3 | 372 | AAH32640.1 | |
| DC306552.1 | 3 | 462 |
More Information
Set Membership: |
|

Genomic Map