Assay Details
Target Gene Details
Entrez Gene ID: | 54876 |
Gene Name: | DDB1 and CUL4 associated factor 16 |
Gene Aliases: |
C4orf30 |
Location: |
Chr.4:17798236-17810761 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCAF16 | NM_017741.3 | 3 | 2327 | NP_060211.3 |
| XM_005248169.3 | 3 | 2315 | XP_005248226.1 | |
| XM_005248171.3 | 2 | 2211 | XP_005248228.1 | |
| XM_006713966.3 | 4 | 2486 | XP_006714029.1 | |
| XM_017008327.1 | 2 | 3780 | XP_016863816.1 | |
| XM_017008328.1 | 2 | 3765 | XP_016863817.1 | |
| AK056116.1 |
More Information
Set Membership: |
|

Genomic Map