Assay Details
Target Gene Details
Entrez Gene ID: | 25977 |
Gene Name: | NECAP endocytosis associated 1 |
Gene Aliases: |
EIEE21 |
Location: |
Chr.12:8082211-8097777 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NECAP1 | NM_015509.3 | 8 | 2208 | NP_056324.2 |
| NR_024260.1 | 7 | 2126 | ||
| AI688397.1 | 1 | 394 | ||
| AK074858.1 | 8 | 2165 | BAC11250.1 | |
| AK074880.1 | 8 | 2178 | BAC11264.1 | |
| AK074923.1 | 8 | 2151 | BAC11296.1 | |
| AK075013.1 | 8 | 2185 | BAC11352.1 | |
| AK094805.1 | 3 | 2750 | ||
| AL050272.1 | 7 | 2052 | CAB43373.2 | |
| BC002888.2 | 8 | 2110 | AAH02888.1 | |
| BC067367.1 | 6 | 1880 | AAH67367.1 | |
| BC084551.1 | 4 | 1719 | AAH84551.1 | |
| BC110876.1 | 8 | 2142 | AAI10877.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428270 | Chr.12:7704223 - 8471391 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map