Assay Details
Target Gene Details
Entrez Gene ID: | 24144 |
Gene Name: | tuftelin interacting protein 11 |
Gene Aliases: |
NTR1, Spp382, TIP39, bK445C9.6, hNtr1 |
Location: |
Chr.22:26491927-26512496 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TFIP11 | NM_001008697.1 | 9 | 1067 | NP_001008697.1 |
| NM_012143.2 | 8 | 990 | NP_036275.1 | |
| XM_011530079.1 | 9 | 1050 | XP_011528381.1 | |
| XM_017028735.1 | 9 | 1208 | XP_016884224.1 | |
| AK025443.1 | 7 | 1114 | ||
| AK025739.1 | 4 | 448 | ||
| AK298303.1 | ||||
| AK299964.1 | ||||
| AL050258.1 | 8 | 1003 | CAB43360.1 | |
| AL080147.1 | 8 | 970 | CAB45740.1 | |
| BC011599.2 | 8 | 967 | AAH11599.1 | |
| BC033080.1 | 8 | 968 | AAH33080.1 | |
| BT007274.1 | 5 | 740 | AAP35938.1 | |
| CT841511.2 | 6 | 842 | CAJ86441.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1310e212 | Chr.22:26495513 - 26502925 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map