Assay Details
Target Gene Details
Entrez Gene ID: | 7358 |
Gene Name: | UDP-glucose 6-dehydrogenase |
Gene Aliases: |
GDH, UDP-GlcDH, UDPGDH, UGD |
Location: |
Chr.4:39498755-39527598 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UGDH | NM_001184700.1 | 11 | 1551 | NP_001171629.1 |
| NM_001184701.1 | 11 | 1583 | NP_001171630.1 | |
| NM_003359.3 | 12 | 1752 | NP_003350.1 | |
| XM_005262667.2 | 12 | 1503 | XP_005262724.1 | |
| AF061016.1 | 12 | 1507 | AAC36095.1 | |
| AJ007702.1 | 12 | 1574 | CAA07609.1 | |
| AK097930.1 | 11 | 1390 | ||
| AK297737.1 | 11 | 1423 | ||
| BC022781.1 | 12 | 1572 | AAH22781.1 | |
| BG194936.1 | 1 | 59 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010189 | Chr.4:39423149 - 39523510 on Build GRCh38 | Gain |
|
| nsv1010784 | Chr.4:39437476 - 39571758 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map