Assay Details
Target Gene Details
Entrez Gene ID: | 254251 |
Gene Name: | ligand dependent nuclear receptor corepressor like |
Gene Aliases: |
MLR1 |
Location: |
Chr.4:17841199-18023287 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LCORL | NM_153686.8 | 7 | 2239 | NP_710153.2 |
| NR_136669.1 | 5 | 2065 | ||
| XM_011513821.2 | 9 | 6970 | XP_011512123.1 | |
| XM_011513822.2 | 8 | 6943 | XP_011512124.1 | |
| XM_011513823.2 | 8 | 6816 | XP_011512125.1 | |
| XM_011513826.2 | 6 | 2023 | XP_011512128.1 | |
| XM_017007961.1 | 8 | 8252 | XP_016863450.1 | |
| XM_017007962.1 | 8 | 6894 | XP_016863451.1 | |
| XM_017007963.1 | 8 | 7197 | XP_016863452.1 | |
| AK096181.1 | 8 | 2100 | ||
| AL133031.1 | 1 | 368 | ||
| BC037322.1 | 7 | 2122 | AAH37322.3 |
Target Gene Details
Entrez Gene ID: | 64151 |
Gene Name: | non-SMC condensin I complex subunit G |
Gene Aliases: |
CAPG, CHCG, NY-MEL-3, YCG1 |
Location: |
Chr.4:17810813-17844865 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCAPG | NM_022346.4 | 21 | 4451 | NP_071741.2 |
| NR_073124.1 | 20 | 4341 | ||
| AK027511.1 | 12 | 2788 | BAB55165.1 | |
| BG398543.1 | 1 | 86 |
More Information
Set Membership: |
|

Genomic Map