Assay Details
Target Gene Details
Entrez Gene ID: | 152503 |
Gene Name: | SH3 domain containing 19 |
Gene Aliases: |
EBP, EVE1, Kryn, SH3P19 |
Location: |
Chr.4:151120281-151228030 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SH3D19 | NM_001009555.3 | 21 | 4472 | NP_001009555.3 |
| NM_001128923.1 | 20 | 4391 | NP_001122395.1 | |
| NM_001128924.1 | 13 | 3394 | NP_001122396.1 | |
| NM_001243349.1 | 20 | 4312 | NP_001230278.1 | |
| XM_005262767.2 | 22 | 4333 | XP_005262824.1 | |
| XM_011531646.1 | 20 | 4119 | XP_011529948.1 | |
| XM_011531647.1 | 19 | 4011 | XP_011529949.1 | |
| XM_011531648.1 | 22 | 4446 | XP_011529950.1 | |
| XM_011531649.1 | 18 | 3942 | XP_011529951.1 | |
| XM_017007778.1 | 22 | 4437 | XP_016863267.1 | |
| XM_017007779.1 | 18 | 3933 | XP_016863268.1 | |
| XM_017007780.1 | 21 | 4368 | XP_016863269.1 | |
| XM_017007781.1 | 21 | 4363 | XP_016863270.1 | |
| XM_017007782.1 | 20 | 4294 | XP_016863271.1 | |
| XM_017007783.1 | 21 | 4264 | XP_016863272.1 | |
| AK128493.1 | 13 | 3243 | ||
| AK294476.1 | 20 | 4312 | ||
| AL133047.1 | 11 | 2236 | CAB61374.1 | |
| BC032468.1 | 3 | 1325 | AAH32468.1 | |
| BC045742.1 | 5 | 1637 | AAH45742.1 | |
| BC085613.1 | 12 | 2451 | ||
| BM831166.1 | ||||
| BX647422.1 | 21 | 4384 | CAI46052.1 | |
| CD675494.1 | 1 | 530 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv595690 | Chr.4:150867803 - 151188400 on Build GRCh38 | Gain |
|
| esv3602674 | Chr.4:151094056 - 151193747 on Build GRCh38 | Gain |
|
| dgv5436n100 | Chr.4:150743286 - 151150724 on Build GRCh38 | Gain |
|
| nsv1028814 | Chr.4:150874356 - 152285445 on Build GRCh38 | Gain |
|
| nsv526976 | Chr.4:150754667 - 151133921 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map