Assay Details
Target Gene Details
Entrez Gene ID: | 383 |
Gene Name: | arginase 1 |
Gene Aliases: |
- |
Location: |
Chr.6:131573159-131584332 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARG1 | NM_000045.3 | NP_000036.2 | ||
| NM_001244438.1 | NP_001231367.1 | |||
| XM_011535801.2 | XP_011534103.1 | |||
| AY074488.1 | AAL71547.1 | |||
| BC005321.1 | AAH05321.1 | |||
| BC020653.1 | AAH20653.1 | |||
| BG217880.1 | ||||
| BT006741.1 | AAP35387.1 | |||
| M14502.1 | AAA51776.1 |
Target Gene Details
Entrez Gene ID: | 9439 |
Gene Name: | mediator complex subunit 23 |
Gene Aliases: |
ARC130, CRSP130, CRSP133, CRSP3, DRIP130, MRT18, SUR-2, SUR2 |
Location: |
Chr.6:131573966-131628239 on Build GRCh38 |
Assay Gene Location: | Within Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MED23 | NM_001270521.1 | 30 | 4511 | NP_001257450.1 |
| NM_015979.3 | 31 | 4529 | NP_057063.2 | |
| AL136776.1 | 31 | 4495 | CAB66710.1 | |
| AW295188.1 | 1 | 102 | ||
| BC060759.1 | 30 | 4455 | AAH60759.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950509 | Chr.6:131570461 - 131576760 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map