Assay Details
Target Gene Details
Entrez Gene ID: | 8545 |
Gene Name: | CGG triplet repeat binding protein 1 |
Gene Aliases: |
CGGBP, p20-CGGBP |
Location: |
Chr.3:88051950-88149866 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CGGBP1 | NM_001008390.1 | 4 | 1176 | NP_001008391.1 |
| NM_001195308.1 | 4 | 1157 | NP_001182237.1 | |
| NM_003663.3 | 3 | 1074 | NP_003654.3 | |
| XM_017007355.1 | 3 | 982 | XP_016862844.1 | |
| AL117392.1 | 3 | 1020 | CAB55894.1 | |
| BC052980.1 | 3 | 976 | AAH52980.1 |
More Information
Set Membership: |
|

Genomic Map