Assay Details
Target Gene Details
Entrez Gene ID: | 1942 |
Gene Name: | ephrin A1 |
Gene Aliases: |
B61, ECKLG, EFL1, EPLG1, LERK-1, LERK1, TNFAIP4 |
Location: |
Chr.1:155127873-155134910 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFNA1 | NM_004428.2 | NP_004419.2 | ||
| NM_182685.1 | NP_872626.1 | |||
| AK057845.1 | ||||
| AK289589.1 | ||||
| AK313117.1 | ||||
| AL833641.1 | ||||
| BC032698.1 | AAH32698.1 | |||
| BC095432.1 | AAH95432.1 | |||
| CR457416.1 | CAG33697.1 | |||
| M57730.1 | AAA58388.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509502 | Chr.1:154904407 - 155142291 on Build GRCh38 | Insertion |
|
| esv3587615 | Chr.1:155047853 - 155175013 on Build GRCh38 | Gain |
|
| nsv998756 | Chr.1:155107614 - 155164069 on Build GRCh38 | Gain |
|
| nsv464084 | Chr.1:155109822 - 155225189 on Build GRCh38 | Loss |
|
| esv2758971 | Chr.1:155122502 - 155345016 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map