Assay Details
Target Gene Details
Entrez Gene ID: | 22921 |
Gene Name: | methionine sulfoxide reductase B2 |
Gene Aliases: |
CBS-1, CBS1, CGI-131, MSRB, PILB |
Location: |
Chr.10:23095498-23122013 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MSRB2 | NM_012228.3 | 5 | 758 | NP_036360.3 |
| AF122004.1 | AAD38899.1 | |||
| AF151889.1 | 5 | 706 | AAD34126.1 | |
| BC018030.1 | AAH18030.1 | |||
| BC117471.1 | ||||
| BC130380.1 |
More Information
Set Membership: |
|

Genomic Map