Assay Details
Target Gene Details
Entrez Gene ID: | 6374 |
Gene Name: | C-X-C motif chemokine ligand 5 |
Gene Aliases: |
ENA-78, SCYB5 |
Location: |
Chr.4:73995642-73998729 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CXCL5 | NM_002994.4 | 4 | 835 | NP_002985.1 |
| BC008376.1 | 4 | 786 | AAH08376.1 | |
| BU169770.1 | ||||
| X78686.1 | 4 | 793 | CAA55355.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1006029 | Chr.4:73928205 - 74120947 on Build GRCh38 | Gain |
|
| esv3575747 | Chr.4:73914851 - 74124725 on Build GRCh38 | Gain |
|
| nsv525980 | Chr.4:73865809 - 74556070 on Build GRCh38 | Gain |
|
| nsv1004768 | Chr.4:73908655 - 74243862 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map