Assay Details
Target Gene Details
Entrez Gene ID: | 201299 |
Gene Name: | RAD52 motif containing 1 |
Gene Aliases: |
RAD52B |
Location: |
Chr.17:35918080-35931821 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RDM1 | NM_001163120.1 | NP_001156592.1 | ||
| NM_001163121.1 | NP_001156593.1 | |||
| NM_001163122.1 | NP_001156594.1 | |||
| NM_001163124.1 | NP_001156596.1 | |||
| NM_001163130.1 | NP_001156602.1 | |||
| NM_145654.3 | NP_663629.1 | |||
| NR_027996.1 | ||||
| NR_027997.1 | ||||
| NR_027998.1 | ||||
| NR_027999.1 | ||||
| NR_028000.1 | ||||
| AB080728.1 | BAC02562.1 | |||
| BX376188.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059795 | Chr.17:35828724 - 36157706 on Build GRCh38 | Gain |
|
| esv3582619 | Chr.17:35887133 - 35918656 on Build GRCh38 | Loss |
|
| nsv960468 | Chr.17:35906705 - 35920915 on Build GRCh38 | Duplication |
|
| nsv2039 | Chr.17:35905026 - 35932726 on Build GRCh38 | Insertion |
|
| esv3892999 | Chr.17:35252863 - 35925533 on Build GRCh38 | Loss |
|
| nsv833424 | Chr.17:35911883 - 36092128 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map