Assay Details
Target Gene Details
Entrez Gene ID: | 4528 |
Gene Name: | mitochondrial translational initiation factor 2 |
Gene Aliases: |
- |
Location: |
Chr.2:55235583-55269311 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTIF2 | NM_001005369.1 | NP_001005369.1 | ||
| NM_001321001.1 | NP_001307930.1 | |||
| NM_001321002.1 | NP_001307931.1 | |||
| NM_001321003.1 | NP_001307932.1 | |||
| NM_001321004.1 | NP_001307933.1 | |||
| NM_001321005.1 | NP_001307934.1 | |||
| NM_002453.2 | NP_002444.2 | |||
| XM_005264335.3 | XP_005264392.1 | |||
| XM_011532871.2 | XP_011531173.1 | |||
| XM_017004162.1 | XP_016859651.1 | |||
| XM_017004163.1 | XP_016859652.1 | |||
| XM_017004164.1 | XP_016859653.1 | |||
| XM_017004165.1 | XP_016859654.1 | |||
| XM_017004166.1 | XP_016859655.1 | |||
| XM_017004167.1 | XP_016859656.1 | |||
| AF494407.1 | AAM14617.1 | |||
| AK225184.1 | ||||
| AK307993.1 | ||||
| BC039848.1 | AAH39848.1 | |||
| BC064977.1 | AAH64977.1 | |||
| L34600.1 | AAA67038.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2720078 | Chr.2:53962225 - 55281329 on Build GRCh38 | Deletion |
|
| nsv522016 | Chr.2:55177658 - 55333162 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map