Assay Details
Target Gene Details
Entrez Gene ID: | 150678 |
Gene Name: | COP9 signalosome subunit 9 |
Gene Aliases: |
CSNAP, MYEOV2 |
Location: |
Chr.2:240126548-240136802 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COPS9 | NM_138336.1 | NP_612209.1 | ||
| XM_017003411.1 | XP_016858900.1 | |||
| AF453951.1 | AAL41026.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997288 | Chr.2:239542278 - 240534009 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115716357] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map