Assay Details
Target Gene Details
Entrez Gene ID: | 221400 |
Gene Name: | tudor domain containing 6 |
Gene Aliases: |
CT41.2, NY-CO-45, SPATA36, TDR2, bA446F17.4 |
Location: |
Chr.6:46687875-46704319 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TDRD6 | NM_001010870.2 | 1 | 3374 | NP_001010870.1 |
| NM_001168359.1 | 1 | 3374 | NP_001161831.1 | |
| AK125838.1 | 1 | 462 | ||
| AK131455.1 | 1 | 3374 | ||
| CR936792.1 | 1 | 1724 |
More Information
Set Membership: |
|

Genomic Map