Assay Details
Target Gene Details
Entrez Gene ID: | 60468 |
Gene Name: | BTB domain and CNC homolog 2 |
Gene Aliases: |
BTBD25 |
Location: |
Chr.6:89926528-90296912 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BACH2 | NM_001170794.1 | 7 | 8118 | NP_001164265.1 |
| NM_021813.3 | 9 | 8289 | NP_068585.1 | |
| XM_005248758.4 | 6 | 9071 | XP_005248815.1 | |
| XM_005248759.4 | 5 | 8991 | XP_005248816.1 | |
| XM_017011156.1 | 12 | 9261 | XP_016866645.1 | |
| XM_017011157.1 | 11 | 9067 | XP_016866646.1 | |
| XM_017011158.1 | 13 | 9340 | XP_016866647.1 | |
| XM_017011159.1 | 14 | 9429 | XP_016866648.1 | |
| XM_017011160.1 | 11 | 9183 | XP_016866649.1 | |
| XM_017011161.1 | 13 | 8891 | XP_016866650.1 | |
| XM_017011162.1 | 10 | 9090 | XP_016866651.1 | |
| XM_017011163.1 | 12 | 9262 | XP_016866652.1 | |
| XM_017011164.1 | 11 | 9168 | XP_016866653.1 | |
| XM_017011166.1 | 10 | 8373 | XP_016866655.1 | |
| AJ271878.1 | 9 | 8123 | CAC28130.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv604125 | Chr.6:89751706 - 90189842 on Build GRCh38 | Gain |
|
| esv3890901 | Chr.6:89204468 - 90033564 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map