Assay Details
Target Gene Details
Entrez Gene ID: | 9358 |
Gene Name: | integrin subunit beta like 1 |
Gene Aliases: |
OSCP, TIED |
Location: |
Chr.13:101452593-101720857 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGBL1 | NM_001271754.1 | 1 | 134 | NP_001258683.1 |
| NM_001271755.1 | 1 | 134 | NP_001258684.1 | |
| NM_004791.2 | 1 | 134 | NP_004782.1 | |
| XM_005254100.4 | 1 | 92 | XP_005254157.1 | |
| AB008375.1 | 1 | 65 | BAA22994.1 | |
| AF072752.1 | 1 | 112 | AAD17666.1 | |
| AK095875.1 | 1 | 97 | ||
| AK291340.1 | ||||
| AK297744.1 | ||||
| AK308194.1 | ||||
| BC036788.1 | AAH36788.2 | |||
| BP284376.1 | 2 | 198 | ||
| DA578691.1 | 1 | 152 | ||
| DA661036.1 | 1 | 276 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892436 | Chr.13:101443165 - 101456121 on Build GRCh38 | Loss |
|
| nsv832701 | Chr.13:101445422 - 101614006 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map