Assay Details
Target Gene Details
Entrez Gene ID: | 27094 |
Gene Name: | potassium calcium-activated channel subfamily M regulatory beta subunit 3 |
Gene Aliases: |
BKBETA3, HBETA3, K(VCA)BETA-3, KCNMB2, KCNMBL, SLO-BETA-3, SLOBETA3 |
Location: |
Chr.3:179239749-179267050 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNMB3 | NM_001163677.1 | NP_001157149.1 | ||
| NM_014407.3 | NP_055222.3 | |||
| NM_171828.2 | NP_741979.1 | |||
| NM_171829.2 | NP_741980.1 | |||
| NM_171830.1 | 1 | 798 | NP_741981.1 | |
| NR_028135.1 | ||||
| AF139471.1 | AAD54771.1 | |||
| AF160968.1 | AAF67811.1 | |||
| AF170916.1 | AAF89698.1 | |||
| AF214561.1 | AAF36598.1 | |||
| AK304837.1 | ||||
| BM745010.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010137 | Chr.3:179245109 - 179332999 on Build GRCh38 | Gain |
|
| esv3893827 | Chr.3:179207079 - 179304740 on Build GRCh38 | Gain |
|
| nsv829806 | Chr.3:179234575 - 179436585 on Build GRCh38 | Gain |
|
| dgv4999n100 | Chr.3:179227018 - 179528391 on Build GRCh38 | Gain |
|
| nsv471116 | Chr.3:179250846 - 179534475 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map