Assay Details
Target Gene Details
Entrez Gene ID: | 27124 |
Gene Name: | inositol polyphosphate-5-phosphatase J |
Gene Aliases: |
INPP5, PIB5PA, PIPP |
Location: |
Chr.22:31107736-31134697 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INPP5J | NM_001002837.2 | NP_001002837.1 | ||
| NM_001284285.1 | NP_001271214.1 | |||
| NM_001284286.1 | NP_001271215.1 | |||
| NM_001284287.1 | NP_001271216.1 | |||
| NM_001284288.1 | NP_001271217.1 | |||
| NM_001284289.1 | NP_001271218.1 | |||
| XM_011530143.1 | XP_011528445.1 | |||
| XM_017028771.1 | XP_016884260.1 | |||
| XM_017028772.1 | XP_016884261.1 | |||
| XM_017028773.1 | XP_016884262.1 | |||
| AK092859.1 | ||||
| AK095944.1 | BAC04657.1 | |||
| AK126610.1 | BAC86611.1 | |||
| AK293992.1 | ||||
| AK308921.1 | ||||
| AK309548.1 | ||||
| BC109288.2 | AAI09289.1 | |||
| BU584792.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829173 | Chr.22:31054057 - 31147970 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map