Assay Details
Target Gene Details
Entrez Gene ID: | 6303 |
Gene Name: | spermidine/spermine N1-acetyltransferase 1 |
Gene Aliases: |
DC21, KFSD, KFSDX, SAT, SSAT, SSAT-1 |
Location: |
Chr.X:23783158-23786223 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SAT1 | NM_002970.3 | 6 | 893 | NP_002961.1 |
| NR_027783.2 | 7 | 1003 | ||
| XM_017029720.1 | 3 | 1521 | XP_016885209.1 | |
| AF251292.1 | 6 | 914 | AAG44592.1 | |
| AL050290.1 | 7 | 964 | ||
| AW874291.1 | 1 | 193 | ||
| BC002503.2 | 6 | 854 | AAH02503.1 | |
| BC008424.1 | 6 | 878 | AAH08424.1 | |
| M77693.1 | 6 | 864 | AAA60573.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470346 | Chr.X:23738748 - 23786754 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map