Assay Details
Target Gene Details
Entrez Gene ID: | 1027 |
Gene Name: | cyclin dependent kinase inhibitor 1B |
Gene Aliases: |
CDKN4, KIP1, MEN1B, MEN4, P27KIP1 |
Location: |
Chr.12:12717270-12722383 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDKN1B | NM_004064.4 | 3 | 1930 | NP_004055.1 |
| AA833962.1 | ||||
| AY004255.1 | 3 | 1512 | AAF88142.1 | |
| BC001971.1 | 3 | 1737 | AAH01971.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2745567 | Chr.12:12246086 - 13092998 on Build GRCh38 | Deletion |
|
| nsv1046355 | Chr.12:11501836 - 12890185 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map