Assay Details
Target Gene Details
Entrez Gene ID: | 65008 |
Gene Name: | mitochondrial ribosomal protein L1 |
Gene Aliases: |
BM022, L1MT, MRP-L1 |
Location: |
Chr.4:77861955-77952790 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL1 | NM_020236.3 | NP_064621.3 | ||
| XM_011532198.2 | XP_011530500.1 | |||
| XM_011532199.1 | XP_011530501.1 | |||
| AB049474.1 | BAB40180.1 | |||
| AF212225.1 | AAF87327.1 | |||
| AK024791.1 | BAB15005.1 | |||
| AK315600.1 | ||||
| BC014356.1 | AAH14356.1 | |||
| BC015109.1 | AAH15109.1 | |||
| BC017765.1 | ||||
| BC032595.1 | AAH32595.1 | |||
| CR457191.1 | CAG33472.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010714 | Chr.4:77821351 - 78069876 on Build GRCh38 | Gain |
|
| nsv516836 | Chr.4:77827823 - 78060765 on Build GRCh38 | Gain |
|
| esv2752050 | Chr.4:77772157 - 78054167 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map