Assay Details
Target Gene Details
Entrez Gene ID: | 25963 |
Gene Name: | transmembrane protein 87A |
Gene Aliases: |
- |
Location: |
Chr.15:42210452-42273584 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM87A | NM_001286487.1 | 19 | 2794 | NP_001273416.1 |
| NM_015497.4 | 20 | 2855 | NP_056312.2 | |
| XM_005254287.1 | 20 | 2709 | XP_005254344.1 | |
| AF129927.1 | 4 | 1191 | ||
| AI434902.1 | 1 | 257 | ||
| AK074870.1 | 20 | 2704 | BAC11256.1 | |
| AL049944.1 | 7 | 1401 | CAB43218.1 | |
| BM989574.1 | 1 | 196 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054102 | Chr.15:42200854 - 42236567 on Build GRCh38 | Gain |
|
| nsv471683 | Chr.15:42042177 - 42210784 on Build GRCh38 | Gain+Loss |
|
| nsv832985 | Chr.15:42079284 - 42272768 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map