Assay Details
Target Gene Details
Entrez Gene ID: | 150962 |
Gene Name: | pseudouridylate synthase 10 |
Gene Aliases: |
CCDC139, DOBI |
Location: |
Chr.2:60938352-61018283 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PUS10 | NM_001322123.1 | 18 | 1837 | NP_001309052.1 |
| NM_001322124.1 | 18 | 1706 | NP_001309053.1 | |
| NM_001322127.1 | 17 | 1832 | NP_001309056.1 | |
| NM_144709.3 | 18 | 1894 | NP_653310.2 | |
| XM_011532568.2 | 19 | 1965 | XP_011530870.1 | |
| XM_011532570.1 | 18 | 1672 | XP_011530872.1 | |
| XM_011532571.1 | 18 | 1846 | XP_011530873.1 | |
| XM_011532572.1 | 18 | 1766 | XP_011530874.1 | |
| XM_011532573.2 | 18 | 1726 | XP_011530875.1 | |
| XM_011532574.2 | 18 | 1787 | XP_011530876.1 | |
| XM_011532576.2 | 18 | 1846 | XP_011530878.1 | |
| XM_011532578.1 | 18 | 1824 | XP_011530880.1 | |
| XM_017003428.1 | 17 | 1663 | XP_016858917.1 | |
| XM_017003429.1 | 17 | 1459 | XP_016858918.1 | |
| AK056874.1 | 18 | 1708 | BAB71300.1 | |
| AK291729.1 | 18 | 1682 | ||
| AL832208.1 | 16 | 4233 | CAI46123.1 | |
| BC101680.1 | 18 | 1686 | AAI01681.1 | |
| BC101706.1 | AAI01707.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834215 | Chr.2:60786854 - 60987072 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map