Assay Details
Target Gene Details
Entrez Gene ID: | 134728 |
Gene Name: | interleukin 1 receptor associated kinase 1 binding protein 1 |
Gene Aliases: |
AIP70, SIMPL |
Location: |
Chr.6:78867486-78979411 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IRAK1BP1 | AK309003.1 |
Target Gene Details
Entrez Gene ID: | 55023 |
Gene Name: | pleckstrin homology domain interacting protein |
Gene Aliases: |
BRWD2, DCAF14, WDR11, ndrp |
Location: |
Chr.6:78934419-79078294 on Build GRCh38 |
Assay Gene Location: | Within Exon 41 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHIP | NM_017934.5 | 39 | 4940 | NP_060404.3 |
| XM_005248729.4 | 39 | 4937 | XP_005248786.1 | |
| XM_011535918.2 | 36 | 4596 | XP_011534220.1 | |
| XM_017010989.1 | 41 | 5082 | XP_016866478.1 | |
| XM_017010990.1 | 38 | 4680 | XP_016866479.1 | |
| AF310250.1 | 16 | 1947 | AAG45145.1 | |
| AJ303102.1 | 39 | 4727 | CAC83118.1 | |
| AK000712.1 | 12 | 1570 | BAA91336.1 | |
| AK075124.1 | 8 | 1027 | BAC11417.1 | |
| AL161957.1 | 16 | 4795 | CAH10776.1 | |
| BC008909.2 | 14 | 1668 | AAH08909.2 | |
| BC081569.1 | 17 | 1983 | AAH81569.1 | |
| BC137488.1 | 39 | 4820 | ||
| DQ924532.1 | 39 | 4714 |
More Information
Set Membership: |
|

Genomic Map