Assay Details
Target Gene Details
Entrez Gene ID: | 84871 |
Gene Name: | ATP/GTP binding protein like 4 |
Gene Aliases: |
CCP6 |
Location: |
Chr.1:48520157-50024011 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGBL4 | NM_001323573.1 | NP_001310502.1 | ||
| NM_001323574.1 | NP_001310503.1 | |||
| NM_001323575.1 | NP_001310504.1 | |||
| NM_032785.3 | NP_116174.3 | |||
| NR_136623.1 | ||||
| XM_011542308.2 | XP_011540610.1 | |||
| XM_017002595.1 | XP_016858084.1 | |||
| XM_017002596.1 | XP_016858085.1 | |||
| XM_017002597.1 | XP_016858086.1 | |||
| AK027348.1 | BAB55053.1 | |||
| AK094826.1 | ||||
| AK294394.1 | ||||
| AK294633.1 | ||||
| AK296221.1 | ||||
| BC126383.1 |
Target Gene Details
Entrez Gene ID: | 79656 |
Gene Name: | BEN domain containing 5 |
Gene Aliases: |
C1orf165 |
Location: |
Chr.1:48727523-48778603 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BEND5 | NM_001302082.1 | 6 | 1407 | NP_001289011.1 |
| NM_024603.3 | 6 | 1325 | NP_078879.2 | |
| XM_011542141.2 | 6 | 3825 | XP_011540443.1 | |
| XM_011542142.2 | 5 | 1273 | XP_011540444.1 | |
| XM_017002331.1 | 7 | 1358 | XP_016857820.1 | |
| XM_017002332.1 | 7 | 1235 | XP_016857821.1 | |
| XM_017002334.1 | 7 | 1454 | XP_016857823.1 | |
| AK021650.1 | 6 | 1269 | BAB13864.1 | |
| AK055449.1 | 4 | 1958 | ||
| BC007932.2 | 6 | 1277 | AAH07932.2 | |
| BC011804.2 | 6 | 1091 | AAH11804.1 | |
| BQ006447.1 | 1 | 111 | ||
| BU743157.1 | 1 | 112 | ||
| BX096297.1 | ||||
| KU178776.1 | 6 | 1187 | ||
| KU178777.1 | 6 | 1269 | ||
| KU178778.1 | 5 | 1053 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829771 | Chr.1:48713186 - 48895189 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map