Assay Details
Target Gene Details
Entrez Gene ID: | 85457 |
Gene Name: | CLOCK interacting pacemaker |
Gene Aliases: |
KIAA1737 |
Location: |
Chr.14:77098235-77117287 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CIPC | NM_033426.2 | 4 | 2464 | NP_219494.2 |
| AB051524.1 | 3 | 2339 | ||
| AK096603.1 | 4 | 2376 | ||
| AL834514.1 | 4 | 2367 | CAD39170.1 | |
| BC037300.2 | 4 | 2384 | AAH37300.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832834 | Chr.14:77088644 - 77258014 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map