Assay Details
Target Gene Details
Entrez Gene ID: | 28512 |
Gene Name: | NFKB inhibitor interacting Ras like 1 |
Gene Aliases: |
KBRAS1, kappaB-Ras1 |
Location: |
Chr.3:23889536-23917046 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKIRAS1 | NM_020345.3 | 5 | 1246 | NP_065078.1 |
| XM_005265076.3 | 5 | 1133 | XP_005265133.1 | |
| XM_005265077.4 | 4 | 1101 | XP_005265134.1 | |
| XM_005265078.3 | 5 | 1085 | XP_005265135.1 | |
| AK027749.1 | 5 | 1223 | BAB55341.1 | |
| BC012145.1 | 5 | 1129 | AAH12145.1 | |
| BC066940.1 | 5 | 1137 | AAH66940.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005186 | Chr.3:21989661 - 24089392 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map