Assay Details
Target Gene Details
Entrez Gene ID: | 58504 |
Gene Name: | Rho GTPase activating protein 22 |
Gene Aliases: |
RhoGAP2, RhoGap22 |
Location: |
Chr.10:48429831-48656265 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP22 | XM_011540013.2 | XP_011538315.1 | ||
| XM_017016473.1 | XP_016871962.1 | |||
| XM_017016476.1 | XP_016871965.1 |
Target Gene Details
Entrez Gene ID: | 5599 |
Gene Name: | mitogen-activated protein kinase 8 |
Gene Aliases: |
JNK, JNK-46, JNK1, JNK1A2, JNK21B1/2, PRKM8, SAPK1, SAPK1c |
Location: |
Chr.10:48306639-48439360 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPK8 | NM_001278547.1 | 12 | 1527 | NP_001265476.1 |
| NM_001278548.1 | 10 | 1121 | NP_001265477.1 | |
| NM_001323302.1 | 14 | 1795 | NP_001310231.1 | |
| NM_001323320.1 | 11 | 1299 | NP_001310249.1 | |
| NM_001323321.1 | 12 | 1532 | NP_001310250.1 | |
| NM_001323322.1 | 14 | 1790 | NP_001310251.1 | |
| NM_001323323.1 | 13 | 1596 | NP_001310252.1 | |
| NM_001323324.1 | 12 | 1532 | NP_001310253.1 | |
| NM_001323325.1 | 13 | 1726 | NP_001310254.1 | |
| NM_001323326.1 | 13 | 1601 | NP_001310255.1 | |
| NM_001323327.1 | 13 | 1726 | NP_001310256.1 | |
| NM_001323328.1 | 13 | 1596 | NP_001310257.1 | |
| NM_001323329.1 | 12 | 1527 | NP_001310258.1 | |
| NM_001323330.1 | 13 | 1721 | NP_001310259.1 | |
| NM_001323331.1 | 13 | 1721 | NP_001310260.1 | |
| NM_139046.3 | 14 | 1795 | NP_620634.1 | |
| NM_139049.3 | 14 | 1790 | NP_620637.1 | |
| NR_136583.1 | 12 | 1528 | ||
| NR_136584.1 | 12 | 1523 | ||
| NR_136585.1 | 12 | 1528 | ||
| AB451271.1 | ||||
| AB451398.1 | ||||
| AK094000.1 | 2 | 1535 | ||
| AK292523.1 | 12 | 1423 | ||
| AL137667.1 | 3 | 2212 | ||
| BC130570.1 | ||||
| BC130572.1 | ||||
| BC144063.1 | ||||
| L26318.1 | 11 | 1318 | AAA36131.1 | |
| U34822.1 | 11 | 1307 | AAC50607.1 | |
| U35004.1 | AAC50610.1 | |||
| U35005.1 | 11 | 1308 | AAC50611.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518744 | Chr.10:48428464 - 48435183 on Build GRCh38 | Loss |
|
| nsv1039508 | Chr.10:48387675 - 48446360 on Build GRCh38 | Gain |
|
| nsv1042677 | Chr.10:47993784 - 49928329 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map