Assay Details
Target Gene Details
Entrez Gene ID: | 2774 |
Gene Name: | G protein subunit alpha L |
Gene Aliases: |
DYT25 |
Location: |
Chr.18:11688956-11885685 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNAL | NM_001142339.2 | 13 | 4967 | NP_001135811.1 |
| NM_001261443.1 | 13 | 4769 | NP_001248372.1 | |
| NM_001261444.1 | 8 | 4487 | NP_001248373.1 | |
| NM_182978.3 | 12 | 5410 | NP_892023.1 |
Target Gene Details
Entrez Gene ID: | 65258 |
Gene Name: | metallophosphoesterase 1 |
Gene Aliases: |
PGAP5 |
Location: |
Chr.18:11883471-11908797 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057995 | Chr.18:10626172 - 11923368 on Build GRCh38 | Gain |
|
| nsv833590 | Chr.18:11757330 - 11924540 on Build GRCh38 | Gain |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map