Assay Details
Target Gene Details
Entrez Gene ID: | 55217 |
Gene Name: | trimethyllysine hydroxylase, epsilon |
Gene Aliases: |
AUTSX6, BBOX2, TMLD, TMLH, TMLHED, XAP130 |
Location: |
Chr.X:155489007-155612961 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMLHE | NM_001184797.1 | 4 | 696 | NP_001171726.1 |
| NM_018196.3 | 4 | 696 | NP_060666.1 | |
| XM_011531182.2 | 4 | 564 | XP_011529484.1 | |
| XM_017029620.1 | 4 | 696 | XP_016885109.1 | |
| AF373407.1 | 4 | 567 | AAL01871.1 | |
| AK001589.1 | 4 | 671 | BAA91775.1 | |
| AK291694.1 | 4 | 634 | ||
| AK304830.1 | 2 | 330 | ||
| AK310667.1 | 2 | 331 | ||
| BC025269.1 | 4 | 612 | AAH25269.1 | |
| CR457265.1 | 3 | 525 | CAG33546.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss |
|
| nsv818043 | Chr.X:155507715 - 155547903 on Build GRCh38 | Gain |
|
| esv29972 | Chr.X:154781072 - 155699751 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map