Assay Details
Target Gene Details
Entrez Gene ID: | 91010 |
Gene Name: | formin like 3 |
Gene Aliases: |
FHOD3, FRL2, WBP-3, WBP3 |
Location: |
Chr.12:49636499-49707414 on Build GRCh38 |
Assay Gene Location: | Within Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FMNL3 | XM_005269218.2 | 27 | 11695 | XP_005269275.1 |
| XM_011538968.2 | 27 | 12185 | XP_011537270.1 | |
| XM_011538969.2 | 26 | 12069 | XP_011537271.1 | |
| XM_011538970.2 | 27 | 12184 | XP_011537272.1 | |
| XM_011538971.2 | 27 | 11760 | XP_011537273.1 | |
| XM_011538972.2 | 26 | 12030 | XP_011537274.1 | |
| XM_011538973.2 | 23 | 11175 | XP_011537275.2 | |
| XM_011538974.2 | 28 | 11517 | XP_011537276.1 |
Target Gene Details
Entrez Gene ID: | 25766 |
Gene Name: | pre-mRNA processing factor 40 homolog B |
Gene Aliases: |
HYPC |
Location: |
Chr.12:49622717-49644669 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRPF40B | NM_001031698.2 | 17 | 1800 | NP_001026868.2 |
| NM_012272.2 | 16 | 2090 | NP_036404.1 | |
| XM_006719324.3 | 16 | 2854 | XP_006719387.1 | |
| XM_006719325.3 | 16 | 2854 | XP_006719388.1 | |
| XM_011538135.2 | 16 | 2854 | XP_011536437.1 | |
| XM_011538136.2 | 16 | 2854 | XP_011536438.1 | |
| XM_011538137.2 | 15 | 2773 | XP_011536439.1 | |
| XM_011538138.2 | 15 | 2743 | XP_011536440.1 | |
| XM_011538139.2 | 17 | 1794 | XP_011536441.1 | |
| XM_011538140.2 | 17 | 1794 | XP_011536442.1 | |
| XM_011538141.1 | 16 | 1620 | XP_011536443.1 | |
| XM_011538143.2 | 16 | 1636 | XP_011536445.1 | |
| XM_011538144.2 | 15 | 1509 | XP_011536446.1 | |
| XM_017019135.1 | 16 | 1557 | XP_016874624.1 | |
| XM_017019136.1 | 15 | 1509 | XP_016874625.1 | |
| XM_017019137.1 | 15 | 1509 | XP_016874626.1 | |
| XM_017019138.1 | 16 | 2854 | XP_016874627.1 | |
| XM_017019139.1 | 12 | 1520 | XP_016874628.1 | |
| AK027117.1 | 2 | 80 | BAB15662.1 | |
| AL137459.1 | 3 | 303 | CAB70747.1 | |
| AL834216.1 | 16 | 2090 | CAD38898.1 | |
| BC050398.2 | 17 | 1636 | AAH50398.1 | |
| BC067364.1 | 16 | 1621 | AAH67364.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832406 | Chr.12:49466910 - 49668353 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map