Assay Details
Target Gene Details
Entrez Gene ID: | 57591 |
Gene Name: | megakaryoblastic leukemia (translocation) 1 |
Gene Aliases: |
BSAC, MAL, MRTF-A |
Location: |
Chr.22:40410281-40636719 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MKL1 | NM_001282660.1 | 12 | 3525 | NP_001269589.1 |
| NM_001282661.1 | 14 | 3787 | NP_001269590.1 | |
| NM_001282662.1 | 15 | 3947 | NP_001269591.1 | |
| NM_001318139.1 | 13 | 3662 | NP_001305068.1 | |
| NM_020831.4 | 15 | 3937 | NP_065882.1 | |
| XM_005261692.1 | 14 | 3742 | XP_005261749.1 | |
| XM_005261694.1 | 14 | 3832 | XP_005261751.1 | |
| XM_011530283.1 | 16 | 4045 | XP_011528585.1 | |
| XM_011530285.1 | 16 | 3956 | XP_011528587.1 | |
| XM_011530287.1 | 8 | 2924 | XP_011528589.1 | |
| XM_017028888.1 | 15 | 4020 | XP_016884377.1 | |
| XM_017028889.1 | 11 | 5020 | XP_016884378.1 | |
| AB037859.2 | 15 | 3909 | ||
| AF368061.1 | 15 | 3862 | AAK54721.1 | |
| AJ297257.1 | 12 | 3519 | CAC38826.1 | |
| AJ297258.1 | 15 | 3919 | CAC38827.1 | |
| AL713710.1 | 7 | 2732 | CAD28507.2 | |
| BC115039.1 | 15 | 3913 | ||
| HG764171.1 | 13 | 3662 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829233 | Chr.22:40404227 - 40438253 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map