Assay Details
Target Gene Details
Entrez Gene ID: | 158399 |
Gene Name: | zinc finger protein 483 |
Gene Aliases: |
ZKSCAN16, ZSCAN48 |
Location: |
Chr.9:111525159-111577844 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF483 | NM_001007169.3 | NP_001007170.1 | ||
| NM_133464.3 | 6 | 1726 | NP_597721.2 | |
| XM_011518300.2 | 6 | 2966 | XP_011516602.1 | |
| XM_017014337.1 | 7 | 1939 | XP_016869826.1 | |
| XM_017014338.1 | 7 | 1825 | XP_016869827.1 | |
| AB075842.1 | 6 | 1652 | ||
| AK092811.1 | BAC03981.1 | |||
| AK291267.1 | 6 | 1718 | ||
| BC136695.1 | 5 | 1519 | ||
| BC136696.1 | 5 | 1519 | ||
| BC152427.1 | 6 | 1652 | ||
| BC152433.1 | 6 | 1652 |
More Information
Set Membership: |
|

Genomic Map