Assay Details
Target Gene Details
Entrez Gene ID: | 8718 |
Gene Name: | TNF receptor superfamily member 25 |
Gene Aliases: |
APO-3, DDR3, DR3, LARD, TNFRSF12, TR3, TRAMP, WSL-1, WSL-LR |
Location: |
Chr.1:6461151-6466195 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFRSF25 | NM_001039664.1 | NP_001034753.1 | ||
| NM_003790.2 | NP_003781.1 | |||
| NM_148965.1 | NP_683866.1 | |||
| NM_148966.1 | NP_683867.1 | |||
| NM_148967.1 | NP_683868.1 | |||
| NM_148970.1 | NP_683871.1 | |||
| AB308321.1 | ||||
| AF026070.1 | AAC39556.1 | |||
| AF026071.1 | AAB82288.1 | |||
| AY358309.1 | AAQ88676.1 | |||
| BC117189.1 | ||||
| BC143886.1 | ||||
| U72763.1 | AAC50819.1 | |||
| U74611.1 | AAB39714.1 | |||
| U75380.1 | AAC51192.1 | |||
| U75381.1 | AAC51193.1 | |||
| U78029.1 | AAB40918.1 | |||
| U83597.1 | AAB41432.1 | |||
| U83598.1 | AAB41433.1 | |||
| U94501.1 | AAC51306.1 | |||
| U94502.1 | AAC51307.1 | |||
| U94503.1 | AAC51308.1 | |||
| U94504.1 | AAC51309.1 | |||
| U94505.1 | AAC51310.1 | |||
| U94506.1 | AAC51311.1 | |||
| U94507.1 | AAC51312.1 | |||
| U94508.1 | AAC51313.1 | |||
| U94509.1 | AAC51314.1 | |||
| U94510.1 | AAC51315.1 | |||
| U94512.1 | AAC51316.1 | |||
| Y09392.1 | CAA70559.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1009548 | Chr.1:6342976 - 6517352 on Build GRCh38 | Loss |
|
| nsv545266 | Chr.1:6464441 - 6470375 on Build GRCh38 | Loss |
|
| dgv2n27 | Chr.1:6396412 - 6512115 on Build GRCh38 | Loss |
|
| nsv954048 | Chr.1:6414241 - 6500740 on Build GRCh38 | Deletion |
|
| nsv7880 | Chr.1:6417968 - 6611914 on Build GRCh38 | Gain+Loss |
|
| nsv470690 | Chr.1:6434339 - 6512115 on Build GRCh38 | Gain |
|
| nsv545264 | Chr.1:6457119 - 6492439 on Build GRCh38 | Loss |
|
| nsv545240 | Chr.1:6351154 - 6512115 on Build GRCh38 | Loss |
|
| nsv470691 | Chr.1:6460251 - 6517705 on Build GRCh38 | Loss |
|
| nsv508836 | Chr.1:6406896 - 6501029 on Build GRCh38 | Insertion |
|
| nsv831536 | Chr.1:6429326 - 6605343 on Build GRCh38 | Loss |
|
| nsv545265 | Chr.1:6463370 - 6520836 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map