Assay Details
Target Gene Details
Entrez Gene ID: | 51742 |
Gene Name: | AT-rich interaction domain 4B |
Gene Aliases: |
BCAA, BRCAA1, RBBP1L1, RBP1L1, SAP180 |
Location: |
Chr.1:235166895-235328240 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARID4B | NM_001206794.1 | 23 | 4139 | NP_001193723.1 |
| NM_016374.5 | 23 | 4260 | NP_057458.4 | |
| NM_031371.3 | 22 | 4002 | NP_112739.2 | |
| NR_135074.1 | 24 | 4418 | ||
| XM_006711781.2 | 23 | 4175 | XP_006711844.1 | |
| XM_011544212.2 | 23 | 4099 | XP_011542514.1 | |
| XM_017001468.1 | 22 | 3921 | XP_016856957.1 | |
| XM_017001469.1 | 22 | 3814 | XP_016856958.1 | |
| XM_017001470.1 | 21 | 3662 | XP_016856959.1 | |
| XM_017001472.1 | 13 | 2933 | XP_016856961.1 | |
| AB210032.1 | 23 | 4139 | ||
| AF083249.1 | 7 | 2123 | AAD41239.1 | |
| AF208045.2 | 21 | 3560 | AAF23433.3 | |
| AF214114.2 | 22 | 3968 | AAF28341.2 | |
| AF227899.1 | 9 | 2445 | AAF36964.1 | |
| AK023144.1 | 4 | 1282 | BAB14428.1 | |
| AL133594.1 | 2 | 325 | CAB63731.1 | |
| AY220790.1 | 21 | 3762 | AAO63590.1 | |
| BC130418.1 | 24 | 4333 |
More Information
Set Membership: |
|

Genomic Map