Assay Details
Target Gene Details
Entrez Gene ID: | 83932 |
Gene Name: | SprT-like N-terminal domain |
Gene Aliases: |
C1orf124, DVC1, PRO4323, spartan |
Location: |
Chr.1:231337936-231355029 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPRTN | NM_001010984.3 | 4 | 3515 | NP_001010984.1 |
| NM_001261462.2 | 3 | 3386 | NP_001248391.1 | |
| NM_032018.6 | 5 | 2477 | NP_114407.3 | |
| XM_006711818.2 | 4 | 2320 | XP_006711881.1 | |
| XM_011544289.2 | 3 | 1803 | XP_011542591.1 | |
| AK027613.1 | 5 | 2157 | BAB55232.1 | |
| AL512744.1 | 5 | 2091 | CAC21670.1 | |
| BC068478.1 | 5 | 2428 | AAH68478.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819245 | Chr.1:231353670 - 231354282 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map