Assay Details
Target Gene Details
Entrez Gene ID: | 139425 |
Gene Name: | DDB1 and CUL4 associated factor 8 like 1 |
Gene Aliases: |
WDR42B |
Location: |
Chr.X:27977993-27981449 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCAF8L1 | NM_001017930.1 | NP_001017930.1 | ||
| AK128123.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6839 | Chr.X:27964445 - 28012816 on Build GRCh38 | Deletion |
|
| nsv518944 | Chr.X:27831821 - 28014899 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map