Assay Details
Target Gene Details
Entrez Gene ID: | 350 |
Gene Name: | apolipoprotein H |
Gene Aliases: |
B2G1, B2GP1, BG |
Location: |
Chr.17:66212029-66229438 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOH | NM_000042.2 | 7 | 894 | NP_000033.2 |
| AK298416.1 | 7 | 653 | ||
| AK313838.1 | 7 | 850 | ||
| BC020703.1 | 7 | 852 | AAH20703.1 | |
| BC026283.1 | 7 | 866 | AAH26283.1 | |
| BM005736.1 | 2 | 249 | ||
| HM212425.1 | 6 | 778 | ||
| M62839.1 | 7 | 874 | AAA51766.1 | |
| S80305.1 | 7 | 844 | AAB21330.1 | |
| X53595.1 | 7 | 868 | CAA37664.1 | |
| X57847.1 | 7 | 868 | CAA40977.1 | |
| X58100.1 | 7 | 856 | CAA41113.1 |
More Information
Set Membership: |
|

Genomic Map