Assay Details
Target Gene Details
Entrez Gene ID: | 55602 |
Gene Name: | CDKN2A interacting protein |
Gene Aliases: |
CARF |
Location: |
Chr.4:183444591-183449064 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDKN2AIP | NM_001317343.1 | 3 | 2230 | NP_001304272.1 |
| NM_017632.3 | 3 | 2261 | NP_060102.1 | |
| AF246705.1 | 3 | 2209 | AAF68967.1 | |
| AK000043.1 | 3 | 2216 | BAA90902.1 | |
| AK096180.1 | 3 | 2097 | ||
| AK225848.1 | ||||
| BC022270.1 | 3 | 2098 | AAH22270.1 | |
| BX538162.1 | 2 | 1734 | CAD98042.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1030766 | Chr.4:183305034 - 183948875 on Build GRCh38 | Gain |
|
| esv26305 | Chr.4:183433937 - 183468540 on Build GRCh38 | Gain |
|
| nsv1033995 | Chr.4:183357552 - 183463120 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map