Assay Details
Target Gene Details
Entrez Gene ID: | 10681 |
Gene Name: | G protein subunit beta 5 |
Gene Aliases: |
GB5 |
Location: |
Chr.15:52120926-52191368 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNB5 | NM_006578.3 | 11 | 1382 | NP_006569.1 |
| NM_016194.3 | 13 | 1487 | NP_057278.2 | |
| XM_011521162.2 | 11 | 1739 | XP_011519464.1 | |
| XM_011521163.2 | 11 | 1264 | XP_011519465.1 | |
| XM_017021867.1 | 8 | 3005 | XP_016877356.1 | |
| AF017656.1 | 11 | 1395 | AAC63826.1 | |
| AK094776.1 | 4 | 1110 | ||
| AK125702.1 | 8 | 1903 | ||
| AL117471.1 | 9 | 1094 | CAB55946.1 | |
| BC013997.2 | 11 | 1379 | AAH13997.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471244 | Chr.15:52013266 - 52219268 on Build GRCh38 | Gain |
|
| nsv833005 | Chr.15:52097672 - 52249133 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map