Assay Details
Target Gene Details
Entrez Gene ID: | 56147 |
Gene Name: | protocadherin alpha 1 |
Gene Aliases: |
PCDH-ALPHA1 |
Location: |
Chr.5:140786136-141012344 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHA1 | NM_018900.3 | NP_061723.1 | ||
| NM_031411.2 | NP_113599.1 | |||
| AF152305.1 | AAD43699.1 | |||
| AF169695.1 | AAF89692.1 |
Target Gene Details
Entrez Gene ID: | 56146 |
Gene Name: | protocadherin alpha 2 |
Gene Aliases: |
PCDH-ALPHA2 |
Location: |
Chr.5:140794859-141012344 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHA2 | NM_018905.2 | NP_061728.1 | ||
| AF152310.1 | AAD43704.1 |
Target Gene Details
Entrez Gene ID: | 56145 |
Gene Name: | protocadherin alpha 3 |
Gene Aliases: |
PCDH-ALPHA3 |
Location: |
Chr.5:140801198-141012344 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHA3 | NM_018906.2 | NP_061729.1 | ||
| AF152311.1 | AAD43705.1 |
Target Gene Details
Entrez Gene ID: | 56144 |
Gene Name: | protocadherin alpha 4 |
Gene Aliases: |
CNR1, CNRN1, CRNR1, PCDH-ALPHA4 |
Location: |
Chr.5:140807074-141012344 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHA4 | NM_018907.3 | NP_061730.1 | ||
| AB208925.1 | BAD92162.1 | |||
| AF152312.1 | AAD43706.1 |
Target Gene Details
Entrez Gene ID: | 56143 |
Gene Name: | protocadherin alpha 5 |
Gene Aliases: |
CNR6, CNRN6, CNRS6, CRNR6, PCDH-ALPHA5 |
Location: |
Chr.5:140821776-141012344 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDHA5 | NM_018908.2 | NP_061731.1 | ||
| NM_031501.1 | NP_113689.1 | |||
| AF152313.1 | AAD43707.1 | |||
| AF152483.1 | AAD43744.1 | |||
| BC033735.1 | 1 | 152 | AAH33735.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv964950 | Chr.5:140821732 - 140822325 on Build GRCh38 | Duplication |
|
| esv2759382 | Chr.5:140777355 - 141449115 on Build GRCh38 | Gain |
|
| nsv830508 | Chr.5:140675388 - 140869073 on Build GRCh38 | Loss |
|
| esv8310 | Chr.5:140807123 - 140841552 on Build GRCh38 | Loss |
|
| dgv970n67 | Chr.5:140784829 - 140843644 on Build GRCh38 | Gain |
|
| esv2659321 | Chr.5:140807351 - 140821938 on Build GRCh38 | Deletion |
|
| nsv1024237 | Chr.5:140810775 - 140852242 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map