Assay Details
Target Gene Details
Entrez Gene ID: | 157570 |
Gene Name: | establishment of sister chromatid cohesion N-acetyltransferase 2 |
Gene Aliases: |
2410004I17Rik, EFO2, RBS |
Location: |
Chr.8:27771949-27812404 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ESCO2 | BC034641.1 | AAH34641.1 |
Target Gene Details
Entrez Gene ID: | 55872 |
Gene Name: | PDZ binding kinase |
Gene Aliases: |
CT84, HEL164, Nori-3, SPK, TOPK |
Location: |
Chr.8:27809621-27838095 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PBK | NM_001278945.1 | 8 | 1650 | NP_001265874.1 |
| NM_018492.3 | 8 | 1675 | NP_060962.2 | |
| XM_006716368.2 | 8 | 1599 | XP_006716431.1 | |
| AB027249.1 | 8 | 1401 | BAA99576.1 | |
| AB027250.1 | 8 | 1381 | BAA99577.1 | |
| AF189722.1 | 8 | 1365 | AAF69107.1 | |
| AF237709.1 | 8 | 1389 | AAF71521.1 | |
| AK027291.1 | 8 | 1380 | BAB55019.1 | |
| BC015191.1 | 8 | 1389 | AAH15191.1 | |
| EU794685.1 | 8 | 1412 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524133 | Chr.8:27776547 - 27858791 on Build GRCh38 | Loss |
|
| nsv519636 | Chr.8:27778100 - 27831788 on Build GRCh38 | Gain |
|
| dgv7153n100 | Chr.8:27780880 - 27831551 on Build GRCh38 | Loss |
|
| nsv1030500 | Chr.8:27806175 - 27821659 on Build GRCh38 | Loss |
|
| nsv1017032 | Chr.8:27767011 - 27821659 on Build GRCh38 | Loss |
|
| dgv7154n100 | Chr.8:27806404 - 27831551 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map