Assay Details
Target Gene Details
Entrez Gene ID: | 54677 |
Gene Name: | carnitine O-octanoyltransferase |
Gene Aliases: |
COT |
Location: |
Chr.7:87345635-87399796 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CROT | NM_001143935.1 | NP_001137407.1 | ||
| NM_021151.3 | NP_066974.2 | |||
| XM_011516337.2 | XP_011514639.1 | |||
| XM_017012370.1 | XP_016867859.1 | |||
| AF073770.1 | AAD41654.1 | |||
| AF168793.1 | AAF03234.1 | |||
| AK127901.1 | ||||
| AK300114.1 | ||||
| AK313487.1 | ||||
| BC039004.1 | AAH39004.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv607725 | Chr.7:87203287 - 87911933 on Build GRCh38 | Gain |
|
| nsv428177 | Chr.7:87228237 - 87404851 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map