Assay Details
Target Gene Details
Entrez Gene ID: | 55055 |
Gene Name: | zwilch kinetochore protein |
Gene Aliases: |
KNTC1AP, hZwilch |
Location: |
Chr.15:66505083-66549485 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZWILCH | NM_001287821.1 | 5 | 753 | NP_001274750.1 |
| NM_001287822.1 | 4 | 657 | NP_001274751.1 | |
| NM_001287823.1 | 4 | 606 | NP_001274752.1 | |
| NM_017975.4 | 5 | 658 | NP_060445.3 | |
| AK000898.1 | 3 | 263 | ||
| AK023175.1 | 5 | 648 | BAB14446.1 | |
| AK122787.1 | 4 | 445 | ||
| AK291048.1 | 4 | 363 | ||
| BC036900.1 | 5 | 404 | AAH36900.1 | |
| BC090041.1 | 5 | 502 | AAH90041.1 | |
| BX640701.1 | 5 | 720 | CAE45821.1 |
More Information
Set Membership: |
|

Genomic Map