Assay Details
Target Gene Details
Entrez Gene ID: | 91875 |
Gene Name: | tetratricopeptide repeat domain 5 |
Gene Aliases: |
Strap |
Location: |
Chr.14:20289131-20305994 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC5 | NM_138376.2 | 8 | 1031 | NP_612385.2 |
| AK074553.1 | 8 | 1014 | BAC11056.1 | |
| BC008647.1 | 8 | 972 | AAH08647.1 | |
| BC030822.1 | 8 | 975 | AAH30822.1 | |
| BC053538.1 | 8 | 979 | AAH53538.1 | |
| BX248066.1 | 8 | 803 | CAD62359.1 | |
| BX640732.1 | 8 | 910 | CAE45848.1 | |
| BX648584.1 | 8 | 986 |
More Information
Additional Information:
For this assay, SNP(s) [rs113096483] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map