Assay Details
Target Gene Details
Entrez Gene ID: | 2206 |
Gene Name: | membrane spanning 4-domains A2 |
Gene Aliases: |
APY, ATOPY, FCER1B, FCERI, IGEL, IGER, IGHER, MS4A1 |
Location: |
Chr.11:60088261-60098467 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MS4A2 | NM_000139.4 | 7 | 1095 | NP_000130.1 |
| NM_001256916.1 | 6 | 960 | NP_001243845.1 | |
| XM_005273846.4 | 8 | 1410 | XP_005273903.1 | |
| XM_011544850.2 | 8 | 1028 | XP_011543152.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv468579 | Chr.11:60046814 - 60207927 on Build GRCh38 | Gain |
|
| dgv201e214 | Chr.11:60084926 - 60104676 on Build GRCh38 | Loss |
|
| esv3626549 | Chr.11:60093431 - 60115836 on Build GRCh38 | Loss |
|
| nsv832173 | Chr.11:60021411 - 60173087 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map