Assay Details
Target Gene Details
Entrez Gene ID: | 9960 |
Gene Name: | ubiquitin specific peptidase 3 |
Gene Aliases: |
SIH003, UBP |
Location: |
Chr.15:63504511-63594640 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP3 | NM_001256702.1 | 14 | 1532 | NP_001243631.1 |
| NM_006537.3 | 15 | 1664 | NP_006528.2 | |
| NR_046341.1 | 16 | 1817 | ||
| NR_046342.1 | 16 | 1780 | ||
| XM_017022763.1 | 17 | 2663 | XP_016878252.1 | |
| XM_017022764.1 | 16 | 2599 | XP_016878253.1 | |
| XM_017022765.1 | 16 | 1805 | XP_016878254.1 | |
| XM_017022766.1 | 17 | 1848 | XP_016878255.1 | |
| XM_017022767.1 | 18 | 2779 | XP_016878256.1 | |
| XM_017022768.1 | 18 | 2853 | XP_016878257.1 | |
| AF073344.1 | 15 | 1531 | AAD42992.1 | |
| AF077040.1 | 14 | 1308 | AAD27773.1 | |
| AK022931.1 | 13 | 1250 | ||
| AK301236.1 | 14 | 1448 | ||
| AK301563.1 | 9 | 963 | ||
| AK307810.1 | 15 | 1423 | ||
| AK309382.1 | 14 | 1485 | ||
| AK314223.1 | 15 | 1583 | ||
| AY461579.1 | 16 | 1693 | AAT37507.1 | |
| BC018113.2 | 15 | 1529 | AAH18113.1 | |
| BC065300.1 | 15 | 1564 | ||
| BC107137.1 | 15 | 1524 | AAI07138.1 | |
| BC107138.1 | 15 | 1524 | AAI07139.1 | |
| BT007269.1 | 15 | 1435 | AAP35933.1 | |
| BX647760.1 | 16 | 1558 |
Target Gene Details
Entrez Gene ID: | 100130855 |
Gene Name: | USP3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:63587710-63600827 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP3-AS1 | NR_034080.1 | |||
| AK093077.1 | ||||
| DB444752.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833034 | Chr.15:63500671 - 63657512 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map